Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N11840

Dodin

technical grade

Synonym(s):

1-Dodecyl-guanidiniumacetat

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H33N3O2
CAS Number:
Molecular Weight:
287.44
Beilstein/REAXYS Number:
6546960
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

technical grade

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-250

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

CC(O)=O.CCCCCCCCCCCCNC(N)=N

InChI

1S/C13H29N3.C2H4O2/c1-2-3-4-5-6-7-8-9-10-11-12-16-13(14)15;1-2(3)4/h2-12H2,1H3,(H4,14,15,16);1H3,(H,3,4)

InChI key

YIKWKLYQRFRGPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

No certificate of analysis offered with technical grade materials.

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Assini et al.
La Medicina del lavoro, 85(4), 321-326 (1994-07-01)
Current pesticide exposure levels can induce a state of sensitization in exposed workers and only in exceptional situations cause severe intoxication. Three cases are reported--one suffering from urticaria/angio-edema, one from asthma and the other from oculo-rhinitis. Only one subject was
J P Cabral
Journal of basic microbiology, 33(4), 219-225 (1993-01-01)
The release of K+ from Pseudomonas syringae cells treated with dodecylguanidine monoacetate (dodine) was followed with a K(+)-selective glass electrode. Treatment of the cells with 5-15 mumol/l dodine resulted in low levels of K+ release, but higher surfactant concentrations caused
J P Cabral
Canadian journal of microbiology, 38(2), 115-123 (1992-02-01)
Treatment of Pseudomonas syringae cells with 50 microM dodecylguanidine monoacetate (dodine) resulted in the rapid degradation and release of RNA and cell lysis. Higher concentrations resulted in a progressive decrease in the intensity of these responses, and the appearance of
[Determination of dodine in selected plant material].
M Kwast
Roczniki Panstwowego Zakladu Higieny, 32(3), 241-244 (1981-01-01)
Mitsutoshi Aoyagi et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 50(2), 58-63 (2009-05-14)
A method for the determination of dodine in agricultural products was developed by using liquid chromatography-electrospray ionization mass spectrometry (LC/ESI-MS). Dodine was extracted with acetonitrile and then acetonitrile-water (7 : 3) from a sample, and re-extracted with ethyl acetate. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service