Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N11686

Dicyclopentadiene

analytical standard

Synonym(s):

4,7-Methano-3a,4,7,7a-tetrahydroindene, Cyclopentadiene dimer, DCPD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12
CAS Number:
Molecular Weight:
132.20
Beilstein/REAXYS Number:
1904092
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 500 mg

manufacturer/tradename

Chem Service, Inc. PS-2033

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

170 °C (lit.)

mp

33 °C (lit.)

density

0.986 g/mL at 25 °C (lit.)

format

neat

SMILES string

C1C=CC2C3CC(C=C3)C12

InChI

1S/C10H12/c1-2-9-7-4-5-8(6-7)10(9)3-1/h1-2,4-5,7-10H,3,6H2/t7-,8+,9?,10?/m0/s1

Inchi Key

HECLRDQVFMWTQS-AFWXGSBKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

89.6 °F - closed cup

flash_point_c

32 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allergic contact dermatitis from unsaturated polyester resin consisting of maleic anhydride, phthalic anhydride, ethylene glycol and dicyclopentadiene.
Keiko Minamoto et al.
Contact dermatitis, 46(1), 62-63 (2002-03-29)
A S Jones et al.
Journal of the Royal Society, Interface, 4(13), 395-403 (2007-01-26)
Self-healing polymers, based on microencapsulated dicyclopentadiene and Grubbs' catalyst embedded in the polymer matrix, are capable of responding to propagating fatigue cracks by autonomic processes that lead to higher endurance limits and life extension, or even the complete arrest of
Jason M Kamphaus et al.
Journal of the Royal Society, Interface, 5(18), 95-103 (2007-06-21)
Using self-healing materials in commercial applications requires healing chemistry that is cost-effective, widely available and tolerant of moderate temperature excursions. We investigate the use of tungsten (VI) chloride as a catalyst precursor for the ring-opening metathesis polymerization of exo-dicyclopentadiene (exo-DCPD)
E N Brown et al.
Journal of microencapsulation, 20(6), 719-730 (2003-11-05)
Microencapsulated healing agents that possess adequate strength, long shelf-life and excellent bonding to the host material are required for self-healing materials. Urea-formaldehyde microcapsules containing dicyclopentadiene were prepared by in situ polymerization in an oil-in-water emulsion that meet these requirements for
L G Stehmeier et al.
Canadian journal of microbiology, 42(10), 1051-1060 (1996-10-01)
Degradation of dicyclopentadiene (DCPD) to carbon dioxide and oxygenated intermediates was established in the laboratory. Screening of many inocula using BIOLOGMT plates showed that no single colony isolate readily mineralized DCPD. Mixed cultures from a variety of environmental sources produced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service