Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N11143

Bentazon methyl derivative

analytical standard

Synonym(s):

N-Methylbentazon

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O3S
CAS Number:
Molecular Weight:
254.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 50 mg

manufacturer/tradename

Chem Service, Inc. PS-1112

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

CC(C)N1C(=O)c2ccccc2N(C)S1(=O)=O

InChI

1S/C11H14N2O3S/c1-8(2)13-11(14)9-6-4-5-7-10(9)12(3)17(13,15)16/h4-8H,1-3H3

Inchi Key

XFTQFXBQDVWOCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service