MSCAL5-10EA
ProteoMass™ LTQ/FT-Hybrid ESI Pos. Mode Cal Mix
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12142208
product line
ProteoMass™
packaging
pkg of 10 × 10 mL
concentration
(varied conc.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
shipped in
dry ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
The ProteoMass™ ESI Calibration Kit for LTQ based Hybrids, positive ion mode calibration solution, enables calibration of Thermo Scientific hybrid instruments equipped with ESI source in positive ion mode and covers a mass range from m/z 138 to m/z 1822 and is therefore useable for calibrations between m/z 50 and 2000.
Features and Benefits
Discover the Advantages for Yourself!
- Ready-to-use
- Developed and qualified specifically for use with the LTQ based Thermo Scientific hybrid instruments (LTQ FT, LTQ FT Ultra, and LTQ Orbitrap series) with ESI source and Thermo Scientific Exactive
- Conveniently packaged, freeing you from time consuming mixing and dilution steps, allowing you to focus on acquisition of quality data
Legal Information
ProteoMass is a trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
ProteoMass is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service