Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MET674A

Supelco

3,5,6-Trichloro-2-pyridinol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. MET-674A

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

InChI

1S/C5H2Cl3NO.Na/c6-2-1-3(7)5(10)9-4(2)8;/h1H,(H,9,10);/q;+1/p-1

InChI key

JAWYJCBOOSFRKD-UHFFFAOYSA-M

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aaron Blair et al.
Occupational and environmental medicine, 68(7), 537-541 (2011-01-25)
The Agricultural Health Study (AHS) is a prospective study of licensed pesticide applicators and their spouses in Iowa and North Carolina. We evaluate the impact of occupational pesticide exposure misclassification on relative risks using data from the cohort and the
Na Wu et al.
Clinical toxicology (Philadelphia, Pa.), 50(6), 514-517 (2012-06-19)
Sodium 3,5,6-trichloropyridin-2-ol (STCP) is a widely used intermediate for the production of chlorpyrifos. However, its effect on the health of workers in STCP factories has become increasingly problematic. This article reports four workers who worked in a STCP factory developed
Melinda Bigelow Dyk et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 46(1), 97-104 (2011-01-18)
Pesticide handlers and pet owners who use products such as shampoos and dips and insecticide-impregnated collars to treat and control fleas on companion animals are exposed to a variety of active ingredients. Chlorpyrifos exposures of adults and children were measured
Berna van Wendel de Joode et al.
Environmental research, 117, 17-26 (2012-07-04)
The US Environmental Protection Agency voluntary phased-out residential use of chlorpyrifos in 2001. In contrast, in Costa Rica, chlorpyrifos-treated bags are increasingly used to protect banana and plantain fruits from insects and to fulfill product standards, even in populated areas.
Li Cao et al.
Current microbiology, 65(3), 231-236 (2012-05-31)
A bacterial strain P2 capable of degrading 3,5,6-trichloro-2-pyridinol (TCP) was isolated and characterized. Phylogenetic analysis based on 16S rRNA gene sequence indicated that it belonged to the genus of Cupriavidus, because it showed the highest sequence similarity to Cupriavidus pauculus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service