Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MET612B

Supelco

Fenamiphos-sulfone

ampule of 100 mg

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22NO5PS
CAS Number:
Molecular Weight:
335.36
Beilstein/REAXYS Number:
2395109
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. MET-612B

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOP(=O)(NC(C)C)Oc1ccc(c(C)c1)S(C)(=O)=O

InChI

1S/C13H22NO5PS/c1-6-18-20(15,14-10(2)3)19-12-7-8-13(11(4)9-12)21(5,16)17/h7-10H,6H2,1-5H3,(H,14,15)

InChI key

LVNYJXIBJFXIRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

replaced by

Product No.
Description
Pricing

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of laboratory lighting on the stability of analytical grade pesticides.
K L Crepeau et al.
Bulletin of environmental contamination and toxicology, 45(3), 446-451 (1990-09-01)
Tanya Cáceres et al.
The Science of the total environment, 398(1-3), 53-59 (2008-05-03)
The acute toxicity of an organophosphorous pesticide, fenamiphos and its metabolites, fenamiphos sulfoxide (FSO), fenamiphos sulfone (FSO(2)), fenamiphos phenol (FP), fenamiphos sulfoxide phenol (FSOP) and fenamiphos sulfone phenol (FSO(2)P), to the aquatic alga Pseudokirchneriella subcapitata and the terrestrial alga Chlorococcum
R Singh
The Analyst, 114(4), 425-427 (1989-04-01)
A method for the simultaneous determination of fenamiphos and its two metabolites, fenamiphos sulphoxide and fenamiphos sulphone, in water is described. The proposed method is based on the separation of the compounds on a silica gel column with ultraviolet detection

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service