Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LOEASTM1

Supelco

LUBRICATING OIL-Ca,P,S,Zn

analytical standard

Synonym(s):

Lubricating Oil w/Elements Standard, Lubricating oil with metals standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

agency

ASTM® D4927,D6443,D6481

packaging

set of 17 × 100 mL

technique(s)

AAS: suitable
ICP: suitable

application(s)

petroleum

format

matrix material

storage temp.

room temp

General description

A number of petroleum based oil products are formulated with organo-metallic additives. These additives function as detergents, antioxidants and/or anti-wear agents. Supelco and Analytical Services (TX) have partnered to provide XRF calibration standards to measure the amount of these additives in newly manufactured oil products, additives and additive packages. Each standard is accompanied with a detailed certificate of analysis (COA).
Same as LOE17, but slightly different concentration ranges. 17 standards per set, 100 mL each.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - STOT SE 3 - Water-react 2

target_organs

Central nervous system

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service