Key Documents
K5161-U
Omnifit® EZ Benchmark Replacement Glass Columns
I.D. × L 15 mm × 150 mm
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
manufacturer/tradename
Omnifit 006ERG-15-15
technique(s)
LPLC: suitable
I.D. × L
15 mm × 150 mm
General description
Replacement glass columns are available in a wide range of bore sizes and lengths. This not only enables you to replace a column should breakage occur, but allows you to obtain several different length columns to use with endpieces you already have for that bore size. The modular nature of our column ranges means that for the ultimate flexibility you can mix and match column sizes with different endpiece types to your specific requirements.
Legal Information
Omnifit is a registered trademark of Diba Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)