K5139-U
Omnifit® EZ Benchmark Columns with 2 Fixed Endpieces
I.D. × L 15 mm × 250 mm
About This Item
manufacturer/tradename
Omnifit 006EZ-15-25-FF
technique(s)
LPLC: suitable
I.D. × L
15 mm × 250 mm
General description
- Non-adjustable with two fixed endpieces (-FF), a cost-effective option when height adjustment is not important.
- One fixed endpiece as a bed support and one adjustable endpiece (plunger) to adjust the bed height (-AF). The adjustable endpiece is designed to give the user fine control over the movement of the endpiece, minimizing the risk of disturbance to the packed bed and giving 80mm of height adjustment.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)