Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K08971338

Kromasil® EternityShell C18 HPLC Column

2.5 μm particle size, L × I.D. 100 mm × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

availability

available only in USA, Canada and Puerto Rico

technique(s)

HPLC: suitable

L × I.D.

100 mm × 2.1 mm

matrix active group

octyldecyl phase

particle size

2.5 μm

Looking for similar products? Visit Product Comparison Guide

General description

The Kromasil EternityShell columns are based on solid-core particles and used in the analysis of pharmaceutical, clinical, environmental, food and beverage and industrial samples. These new columns are packed with 2.5 μm solid-core particles. Kromasil EternityShell are aimed for fast separations similarly to the already existent Kromasil EternityXT columns with 1.8 and 2.5 μm particles.

Extended pH range

The Kromasil EternityShell C18 columns are robust and stable beyond traditional silica columns. Offering excellent pH flexibility between pH 1 and 12, These EternityShell columns are for users that need to work under an almost full pH range, including acidic conditions but also at tougher high pH for the analysis for bases. The new chromatographic material is endcapped, has a surface area of 110 m2/g and a carbon coverage of 8%, providing a first rate alternative selectivity for reversed phase chromatography.

Legal Information

Kromasil is a registered trademark of EKA Chemicals AB

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service