Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K08971335

Kromasil® ClassicShell C18 HPLC Column

2.5 μm particle size, L × I.D. 100 mm × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NB.21

product line

Kromasil®

manufacturer/tradename

Kromasil®

availability

available only in USA, Canada and Puerto Rico

technique(s)

HPLC: suitable

L × I.D.

100 mm × 2.1 mm

matrix active group

C18 (octadecyl) phase

particle size

2.5 μm

Looking for similar products? Visit Product Comparison Guide

General description

The Kromasil ClassicShell columns are based on solid core particles and intended for the analysis of sample mixtures in various areas of research as well as quality control in pharmaceutical, environmental, food and beverages and industrial laboratories.

These new columns are packed with 2.5 μm particles and, similarly to the already adopted Kromasil Classic 1.8 and 2.5 μm particle size columns, Kromasil ClassicShell products are intended for fast separations to support effective and efficient laboratory turnaround.

Legal Information

Kromasil is a registered trademark of EKA Chemicals AB

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service