K08971148
Kromasil® C8, Bulk
10 μm particle size, pore size 100 Å, pkg of 1000 g
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115700
NACRES:
NB.21
product line
Kromasil®
packaging
pkg of 1000 g
manufacturer/tradename
Kromasil® KR100-10-C8-1000GM
availability
available only in USA, Canada and Puerto Rico
matrix active group
C8 (octyl) phase
particle size
10 μm
pore size
100 Å pore size
Looking for similar products? Visit Product Comparison Guide
Legal Information
Kromasil is a registered trademark of EKA Chemicals AB
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service