Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IP336LMPSE

Polysulfide in light mineral oil, IP 336

Calibration Set E

Sign Into View Organizational & Contract Pricing

Select a Size

7 X 100 ML
$928.00

$928.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
7 X 100 ML
$928.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$928.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

Looking for similar products? Visit Product Comparison Guide

Components

Kit contains: Blank, LMPS070, LMPS080,LMPS090, LMPS110, LMPS130, LMPS140. 100 mL ea

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service