Key Documents
I263330
Sigma-Aldrich
Inlet Liner, Split, for Shimadzu SPL-14 injector
About This Item
UNSPSC Code:
12352200
packaging
pkg of 1 ea
manufacturer/tradename
Shimadzu 221-32544-01 (OEM)
liner L × O.D.
99 mm × 5 mm
compatibility
configured for
for use with Shimadzu 14/15A/16 with SPL-14 Injectors
technique used for General purpose
Other Notes
I263330 --> material nos. 2633305 (5pk) & 2633325 (25pk).
Legal Information
Shimadzu is a trademark of Shimadzu Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)