Key Documents
I263100
Inlet Liner, packed, 4mm ID, for PerkinElmer® Split/Splitless injector
About This Item
UNSPSC Code:
12352200
Pricing and availability is not currently available.
description
packed (deactivated glass wool)
packaging
pkg of 1 ea
liner L × O.D. × I.D.
92 mm × 1/4 in. × 4 mm
compatibility
configured for
for use with PerkinElmer Auto System, Model 9000 Split/Splitless Capillary Injector
technique used for General purpose; Dirty samples
Other Notes
I263100 (I263090+I203650) --> material nos. 2631005 (5pk) & 2631025 (25pk).
Legal Information
PerkinElmer is a registered trademark of PerkinElmer Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)