FFA57302
SPME Fiber Assembly
df 7 μm(PDMS, for use with multi fiber exchanger, needle size 24 ga
About This Item
material
fused silica fiber
plain green hub
needle size
24 ga
packaging
pkg of 3 × ea
df
7 μm (PDMS)
technique(s)
solid phase microextraction (SPME): suitable
fiber L
1 cm
matrix active group
PDMS coating
application(s)
food and beverages
compatibility
for analyte group volatiles (MW 80-500)
for use with multi fiber exchanger
Looking for similar products? Visit Product Comparison Guide
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
SPME fast-fit fiber assemblies simplify method development for solid-phase microextraction with easy installation and color-coded labels.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service