FFA57295-U
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS)
df 85 μm(CAR/PDMS, needle size 23 ga, Stableflex, for use with multi fiber exchanger
About This Item
Recommended Products
material
fused silica SPME fiber
plain light blue hub
needle size
23 ga
packaging
pkg of 3 ea
df
85 μm (CAR/PDMS)
technique(s)
solid phase microextraction (SPME): suitable
fiber L
1 cm
matrix active group
CAR/PDMS coating
application(s)
food and beverages
compatibility
(For gasses and low molecular weight compounds (MW 30 - 225).)
for use with multi fiber exchanger
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
SPME fast-fit fiber assemblies simplify method development for solid-phase microextraction with easy installation and color-coded labels.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service