Kit 1 - For Volatiles and Semivolatiles - contains one fiber of each:
85μm polyacrylate coating
100μm polydimethylsiloxane coating
7μm polydimethylsiloxane coating
Other Notes
Although we no longer distribute SPME-FFA products andaccessories, these products are available and distributed exclusively byChromline. Please visit the Chromline websiteor contact [email protected] for more information.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
International journal of legal medicine, 109(3), 150-154 (1996-01-01)
The most common alkyl nitrites encountered in forensic toxicology are iso-butyl, n-butyl and iso-pentyl(amyl) nitrites. All have become popular as an aphrodisiac, especially among the homosexual population. Alkyl nitrites are a volatile and unstable group of compounds, which hydrolyse in
Journal of separation science, 35(19), 2544-2552 (2012-08-22)
The presence of triclosan and triclocarban, two endocrine-disrupting chemicals and antimicrobial agents, and transformation products of triclocarban, 1,3-di(phenyl)urea, 1,3-bis(4-chlorophenyl)urea and 1,3-bis(3,4-dichlorophenyl)urea, in tap water, treated household drinking water, bottled water, and river water samples were investigated using solid-phase micro-extraction coupled
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(12), 1832-1839 (2012-08-23)
Selected commercial processed foods available in the Brazilian market (306 samples) were analysed for furan content using a validated gas chromatography-mass spectrometry method preceded by headspace solid phase micro-extraction (HS-SPME-GC/MS). Canned and jarred foods, including vegetable, meat, fruit and sweet
2-Methoxy-3-isobutylpyrazine (MIBP) contributes a bell pepper aroma to many grape cultivars and has a reported aroma threshold of ∼2 ng L(-1) in water. The purpose of this study was twofold: (1) develop a procedure using headspace solid phase micro-extraction combined
Journal of food science, 73(8), E349-E355 (2008-11-21)
Physical properties of whey protein isolate (WPI) coating solution incorporating ascorbic palmitate (AP) and alpha-tocopherol (tocopherol) were characterized, and the antioxidant activity of dried WPI coatings against lipid oxidation in roasted peanuts were investigated. The AP and tocopherol were mixed
SPME fast-fit fiber assemblies simplify method development for solid-phase microextraction with easy installation and color-coded labels.
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.