Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FAP88819

Nutritional elements in milk powder

reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701

grade

reference material

manufacturer/tradename

Fera Science Ltd

application(s)

food and beverages

format

matrix material

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Trace Elements
Calcium (Ca), Iodine (I), Magnesium (Mg), Phosphorous (P), Potassium (K), Selenium (Se), Sodium (Na), Iron (Fe), Copper (Cu), Zinc (Zn), Manganese (Mn), Molybdenum (Mo)

Matrix Group: Dairy products and eggs

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service