FAP88659
Nutritional elements in infant formula
reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
85151701
Recommended Products
grade
reference material
manufacturer/tradename
Fera Science Ltd
application(s)
food and beverages
format
matrix material
storage temp.
−20°C
Other Notes
Certified for the analytes listed below. See certificate for values and more details
Trace Elements
Calcium (Ca), Copper (Cu), Iodine (I), Iron (Fe), Magnesium (Mg), Manganese (Mn), Phosphorous (P), Potassium (K), Selenium (Se), Sodium (Na), Zinc (Zn)
Matrix Group: Dairy products and eggs
Trace Elements
Calcium (Ca), Copper (Cu), Iodine (I), Iron (Fe), Magnesium (Mg), Manganese (Mn), Phosphorous (P), Potassium (K), Selenium (Se), Sodium (Na), Zinc (Zn)
Matrix Group: Dairy products and eggs
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service