Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FAP80679

Soft drinks ingredients

reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$295.00

$295.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 EA
$295.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

$295.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

grade

reference material

Quality Level

manufacturer/tradename

Fera Science Ltd

application(s)

food and beverages

format

matrix material

storage temp.

−20°C

Related Categories

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals
Benzoic acid, Caffeine
Sweeteners
Acesulfame-K, Saccharin

Matrix Group: Beverages and Water

pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT RE 2 Inhalation

target_organs

Lungs

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service