The concentration of the Analytes are listed on the lot specific Certificate of Analysis. Please see the sample Certificate below
https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/PDF737307.pdf
Recommended Products
grade
reference material
packaging
pkg of 150 g
manufacturer/tradename
Fera Science Ltd
application(s)
food and beverages
format
matrix material
Other Notes
Trace Elements
Arsenic (As), Cadmium (Cd), Mercury (Hg)
Matrix Group: Fish and Seafood
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the analyte concentration of this product?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service