Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ES5411

ChromegaChiral Chiral CCA, 5 Micron HPLC Column

5 μm particle size, L × I.D. 25 cm × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.52

material

stainless steel column

product line

ChromegaChiral

manufacturer/tradename

ES Industries 152251-CCA

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

25 cm × 2.1 mm

matrix

fully porous particle

matrix active group

carbamoyl phase

particle size

5 μm

pore size

1000 Å pore size

operating pH

2-8

separation technique

chiral
supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

A polysaccharide coated chiral stationary phase and columns which are produced using a unique production process of coating the proven chiral selector, tris-(3,5-di- methylphenyl) carbamoyl amylose on high purity silica gel. ChromegaChiral CCA columns, similar in selectivity to ChiralPak® AD, are available in 3, 5, 10, and 20 micron particle sizes enabling easy scale up from analytical to preparative scale using HPLC or SFC conditions.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

CHIRALPAK is a registered trademark of Daicel Corp.
ChromegaChiral is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service