Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ES5238

GreenSep Ethyl Pyridine II SFC 3 Micron HPLC Column

3 μm particle size, L × I.D. 15 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.52

material

stainless steel column

product line

GreenSep

manufacturer/tradename

ES Industries 135191-GS-PYE-II

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

fully porous particle

matrix active group

2-ethyl pyridine phase

particle size

3 μm

pore size

120 Å pore size

operating pH

2-8

separation technique

supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

The column ideally suited for the retention and rapid separation of chemicals containing acidic groups. The chromatogram below shows the separation of chemicals that contain acidic groups and demonstrates the retention and separation capability that GreenSep Ethyl Pyridine II can deliver for the SFC chromatographer.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service