Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ES5165

GreenSep DEAP SFC 3 Micron HPLC Column

3 μm particle size, L × I.D. 5 cm × 3.0 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.52

material

stainless steel column

product line

GreenSep

manufacturer/tradename

ES Industries 113191-GS-DEAP

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

5 cm × 3.0 mm

matrix

fully porous particle

matrix active group

DEAP phase

particle size

3 μm

pore size

120 Å pore size

operating pH

2-8

separation technique

supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

Unlike many SFC columns which require the addition of amine modifiers to the mobile phase to improve peak shape for basic compounds, GreenSep DEAP does not, greatly simplifying mobile phase composition and fraction collection. This makes GreenSep DEAP the SFC column of choice for the retention and rapid separation of chemical compounds containing strong amine groups.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service