Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ES5015

GreenSep Basic SFC 1.8 Micron HPLC Column

1.8 μm particle size, L × I.D. 10 cm × 3.0 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.52

material

stainless steel column

product line

GreenSep

manufacturer/tradename

ES Industries 523A91-GS-BC

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

10 cm × 3.0 mm

matrix

fully porous particle

matrix active group

amine phase

particle size

1.8 μm

pore size

120 Å pore size

operating pH

2-8

separation technique

supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

GreenSep Basic exhibits a highly basic character and is ideally suited to high performance/high speed SFC preparative chromatography applications of chemicals containing amine groups. The chromatogram shown below contains chemicals that are functionalized with amine and acidic groups and is a prime example of the superior peak shape performance and separation capacity obtainable with the GreenSep Basic column. It also compares GreenSep Basic with GreenSep Ethyl Pyridine showing the superior reten- tion on GreenSep Basic when compared to GreenSep Ethyl Pyridine.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service