Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ERMBD514

Cd in Cocoa

BAM Certified Reference Material, Cd (Medium)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

manufacturer/tradename

BAM

concentration

Cd (Medium)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service