Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMB2278

Rice Flour (C, H, N, S)

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

manufacturer/tradename

Elemental Microanalysis Limited

application(s)

food and beverages

format

matrix material

General description

Rice Flour (C, H, N, S), under the category of organic standards from elemental microanalysis, are traceable to the national institute of standards and technology (N.I.S.T.) Maryland USA and/or (where appropriate) other accredited certification organizations. These materials are suitable for use as working standards for elemental analysis and are supplied with batch certificates of analysis.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Elemental Analysis: Carbon, Hydrogen, Nitrogen and Sulphur

Matrix Group: Cereals, Bread, Rice and Maize

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service