Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMB2140

Pasta (Nitrogen)

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

manufacturer/tradename

Elemental Microanalysis Limited

application(s)

food and beverages

format

matrix material

General description

Pasta (Nitrogen), an organic analytical standard (OAS), consists of a homogeneous batch of pasta for use as a routine working microanalytical standard for the determination of nitrogen. This OAS should be stored at temperatures between 20°C to 25°C and should be kept tightly sealed away from the light and moisture. It is non-hygroscopic under normal conditions and can be used without preliminary drying.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Elemental Analysis: Nitrogen

Matrix Group: Cereals, Bread, Rice and Maize

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service