EC10A
Fatty acids, saturated, even-carbon
99-100% (GLC, TLC)
About This Item
Recommended Products
assay
99-100% (GLC, TLC)
Storage temp.
-10 to -25°C
Application
Other Notes
C 2250 Caproic Acid (C6:0), 10 ml
C 2875 Caprylic Acid (C8:0), 10 ml
C 1875 Capric Acid (C10:0), 10 g
L 4250 Lauric Acid (C12:0), 10 g
M 3128 Myristic Acid (C14:0), 10 g
P 0500 Palmitic Acid (C16:0), 10 g
S 4751 Stearic Acid (C18:0), 5 g
A 3631 Arachidic Acid (C20:0), 5 g
B 7644 Behenic Acid (C22:0), 5 g
L 6641 Lignoceric Acid (c24:0), 1 g
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service