Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM44898

Supelco

ASTM D6584 Individual Stock and Internal StandardsMix Kit

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

certified reference material
TraceCERT®

agency

ASTM® D6584

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

kit of

technique(s)

gas chromatography (GC): suitable

format

single component solution

storage temp.

2-8°C

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials

Kit Components Also Available Separately

Product No.
Description
SDS

  • CRM44895ASTM® D6584 Triolein solution, certified reference material, 5000 μg/mL in pyridine, ampule of 1 × 2 mLSDS

  • CRM44894ASTM® D6584 1,3-Diolein solution, certified reference material, 5000 μg/mL in pyridineSDS

  • CRM44896ASTM® D6584 1,2,4-Butanetriol Solution, certified reference material, 1000 μg/mL in pyridine, ampule of 5 mLSDS

  • CRM44897ASTM® D6584 Tricaprin Solution, certified reference material, 8000 μg/mL in pyridineSDS

  • CRM44892ASTM® D6584 Glycerin solution, certified reference material, 500 μg/mL in pyridineSDS

  • CRM44893ASTM® D6584 Monoolein solution, certified reference material, 5000 μg/mL in pyridineSDS

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service