Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM42222

GC/MS Pesticides Calibration Mix 9

certified reference material, 100 μg/mL each component in acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

product line

TraceCERT®

packaging

ampule of 1 mL

concentration

100 μg/mL each component in acetonitrile

technique(s)

GC/MS: suitable

format

multi-component solution

storage temp.

−20°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Azaconazole 100 μg/mL
    Bitertanol 100 μg/mL
    Bromuconazole 100 μg/mL
    Butralin 100 μg/mL
    Cadusafos 100 μg/mL
    Cloquintocet-mexyl 100 μg/mL
    Cyprazine 100 μg/mL
    Cyproconazol 100 μg/mL
    Difenoconazol 100 μg/mL
    Dimepiperate 100 μg/mL
    Dimoxystrobin 100 μg/mL
    Diniconazole 100 μg/mL
    Dipropetryn 100 μg/mL
    Ditalimfos 100 μg/mL
    Etofenprox 100 μg/mL
    Fenamiphos 100 μg/mL
    Fenhexamid 100 μg/mL
    Flamprop-methyl 100 μg/mL
    Fluazinam 100 μg/mL
    Flutriafol 100 μg/mL
    Formothion 100 μg/mL
    Fosthiazate 100 μg/mL
    Indoxacarb 100 μg/mL
    Isocarbophos 100 μg/mL
    Isodrine 100 μg/mL
    Mepanipyrim 100 μg/mL
    Ofurace 100 μg/mL
    Paclobutrazol 100 μg/mL
    Picolinafen 100 μg/mL
    Pyridafenthion 100 μg/mL
    Pyrifenox 100 μg/mL
    Pyrimethanil 100 μg/mL
    Quinoxyfen 100 μg/mL
    Quizalofop-ethyl 100 μg/mL
    Tebufenpyrad 100 μg/mL
    Tetraconazole 100 μg/mL
    Thiobencarb 100 μg/mL
    Triallat 100 μg/mL
    Mexacarbate 100 μg/mL
    See All (39)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service