CRM42205
GC/MS Pesticides Calibration Mix 4
certified reference material, 100 μg/mL each component in acetonitrile
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
grade
certified reference material
TraceCERT®
product line
TraceCERT®
packaging
ampule of
concentration
100 μg/mL each component in acetonitrile
technique(s)
gas chromatography (GC): suitable
format
multi-component solution
storage temp.
−20°C
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
Analyte
Description
Ethyl pirimiphos
Pentachloroaniline
Pentachloronitrobenzene
Phenthoate
Phorate sulfone
Pirimicarb
Pirimiphos-methyl
Procymidone
Profenfos
Propanil
Propham
Pyrazophos
Quinalfos
Safrotin
Tebuconazole
Terbumeton
Terbuthylazine
1,2,4,5-Tetrachloro-3-nitroben
Tetramethrin (mix of isomers)
Tetrasul
Tilt
Tolban
Tolclofos-methyl
Tolylfluanid
Triadimenol
Triazophos
Triflumizole
Vinclozolin
See All (28)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service