Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM42203

Supelco

GC/MS Pesticides Calibration Mix 3

certified reference material, 100 μg/mL each component in acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

packaging

ampule of 1 mL

concentration

100 μg/mL each component in acetonitrile

technique(s)

gas chromatography (GC): suitable

application(s)

agriculture

format

multi-component solution

storage temp.

−20°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Aldrin
    α-1,2,3,4,5,6-Hexachlorocyclohexane
    α-Chlordane
    β-BHC
    Chlorbensid
    Chlorfenson
    Chlorobenzilate
    Chloroneb
    cis-Nonachlor
    Dacthal
    4,4′-DDD
    2,4′-DDD
    2,4′-DDE
    4,4′-DDE
    2,4′-DDT
    4,4′-DDT
    δ-BHC
    Dieldrin
    Endosulfan ether
    α-Endosulfan
    β-Endosulfan
    Endosulfan sulfate
    Endrin
    Endrin aldehyde
    Endrin ketone
    Fenson
    Lindane
    γ-Chlordane
    Heptachlor
    Heptachlor epoxide isomer B
    Methoxychlor
    Permethrin (cis/trans)
    Perthane
    Tetradifon
    trans-Nonaclor
    See All (35)

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service