Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM40471

Supelco

Isoborneol solution

certified reference material, 2000 μg/mL in methanol, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable

application(s)

cannabis testing
cannabis testing

format

single component solution

storage temp.

2-8°C

InChI

1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8+,10-/m0/s1

InChI key

DTGKSKDOIYIVQL-XKSSXDPKSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service