Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Formaldehyde-2,4-DNPH Solution

100 μg/mL in acetonitrile (as aldehyde), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
210.15
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

CofA

certificate of analysis is enclosed in each package.

feature

standard type calibration

packaging

ampule of 5 × 1 mL (4M7177)

concentration

100 μg/mL in acetonitrile (as aldehyde)

storage temp.

2-8°C

InChI

1S/C7H6N4O4/c1-8-9-6-3-2-5(10(12)13)4-7(6)11(14)15/h2-4,9H,1H2

InChI key

UEQLSLWCHGLSML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

These products are now available as certified reference materials. Please reference CRM47177 and CRM4M7177.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service