Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Closures for Snap Ring vials

green polypropylene seal, seal diam. × thickness 11 mm × 1.0 mm, red PTFE/silicone/red PTFE septum

Sign Into View Organizational & Contract Pricing


About This Item

material

green polypropylene seal
red PTFE/silicone/red PTFE septum

packaging

pkg of 100 ea (28857-U)
pkg of 1000 ea (28853-U)

O.D. × H × I.D.

11.5 mm × 6 mm × 9 mm

seal diam. × thickness

11 mm × 1.0 mm

fitting

for 11 mm snap ring

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service