Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EPA 8270 Acids Surrogate Spike Mix HC

analytical standard, 10,000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

grade

analytical standard

agency

EPA 8270

CofA

certificate of analysis is enclosed in each package.

feature

standard type surrogate

packaging

ampule of 4 × 5 mL

concentration

10,000 μg/mL in methanol

application(s)

environmental

storage temp.

2-8°C

Analyte

Description

    2-Fluorophenol
    Phenol-d6
    2,4,6-Tribromophenol

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service