Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Trifluoroacetic acid

Synonym(s):

trifluoroacetic acid, trifluoroacetic anhydride (TFAA), TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein/REAXYS Number:
742035
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

grade

derivatization grade ((GC derivatization))

vapor density

3.9 (vs air)

vapor pressure

97.5 mmHg ( 20 °C)

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

packaging

pkg of 1 × 25 mL (33075)

refractive index

n20/D 1.3 (lit.)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

density

1.489 g/mL at 20 °C (lit.)

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for the derivatization of aminoalkylphosphoric acids.

Other Notes

Reagent for trifluoroacetyl, butyl ester.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service