Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Hydrogen chloride solution

0.5 N in methanol

Synonym(s):

Methanolic HCl, Hydrochloric acid solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
UNSPSC Code:
12352301
grade:
derivatization grade ((GC derivatization))
Pricing and availability is not currently available.

Product Name

Methanolic HCl, 0.5 N

grade

derivatization grade ((GC derivatization))

form

liquid

reaction suitability

reagent type: derivatization reagent
reaction type: Esterifications

packaging

pkg of 10 × 5 mL (33095)

concentration

0.5 N

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanolic HCl (hydrochloric acid in methanol) is particularly useful for preparing methyl esters of volatile (short chain) fatty acids. Fatty acids are esterified by heating them with an anhydrous alcohol (e.g., methanol) in the presence of an acidic catalyst (e.g., HCl) in a sealed vessel at a high temperature for a short time. In the reaction, a fatty acid molecule and an alcohol molecule are joined, with the release of a water molecule. The derivatives can be quickly and easily recovered, quantitatively, from the reaction medium.

Application

Suitable for derivatization of fatty acids, lipids and formic and acetic acid (in cigarett smoke).

Features and Benefits

  • Derivatization of fatty acids, particularly volatile (short chain) fatty acids.

  • Clean reaction (no side reaction) with volatile by-products.

  • Provides clean, fast, quantitative derivatization.

Other Notes

Reagent for methyl esters.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service