Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HMDS+TMCS+Pyridine, 3:1:9 (Sylon HTP)

Synonym(s):

BSA, BSTFA, HMDS, HMDS + TMCS in pyridine, TMCS, pyridine, 1,1,1,3,3,3–Hexamethyldisilane+Τrimethylchlorosilane+Pyridine, Sylon HTP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

grade

derivatization grade ((GC derivatization))

description

3:1:9 (SYLON HTP)

composition

1,1,3,3,3-Hexamethyldisilazane, 22.9%
Pyridine, 69.4%
Trimethylchlorosilane, 7.7 % (w/v)

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 20 × 1 mL (33038)

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for the derivatization of 2-amino-2-dexoxyhexoses, amino sugars, amino suganrs (in polysaccharides), ceramides, cholanthrene dihydrodiol, hexuronic acids, inositols, ketosteroids, monoacyl and monalkyl glycerols, natural glycoses, oligosaccharides, pyridoxine-5-phosphate, spingosines, sugar and sugar phosphates.

Other Notes

Reagent for dihydro, trimethylsilyl, trimethylsilyl (alditol), trimethylsilyl ether and o-trimethylsilyl-aldono-1,4-lactones.

Legal Information

Sylon is a trademark of Sigma-Aldrich Co. LLC
Sylon CT is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service