Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BP956

Aminoglutethimide impurity D

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

3,3′-(Azodi-4,1-phenylene)bis[3-ethyl-2,6-piperidinedione], 3,3′-[Diazenediylbis(4,1-phenylene)]bis(3-ethylpiperidine-2,6-dione), Azoglutethimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28N4O4
CAS Number:
Molecular Weight:
460.52
UNSPSC Code:
41116107

API family

aminoglutethimide

form

solid

manufacturer/tradename

BP

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 10 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service