Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BP678

Fosfestrol sodium

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Fosfestrol tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18O8Na4P2
CAS Number:
Molecular Weight:
516.24
UNSPSC Code:
12352200
Pricing and availability is not currently available.

form

solid

storage temp.

2-8°C

InChI

1S/C18H22O8P2.4Na/c1-3-17(13-5-9-15(10-6-13)25-27(19,20)21)18(4-2)14-7-11-16(12-8-14)26-28(22,23)24;;;;/h5-12H,3-4H2,1-2H3,(H2,19,20,21)(H2,22,23,24);;;;/q;4*+1/p-4/b18-17+;;;;

InChI key

XGZAXRQNRRXUMY-MJCKVQKWSA-J

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

Unit quantity: 50 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service