Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AR2027

Supelco

Barley Flour (C, H, N, S)

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

manufacturer/tradename

Alpha Resources

application(s)

food and beverages

format

matrix material

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Crude Fiber, Crude Fat, Ash (AOAC 942.05), Moisture (AOAC 930.15, 967.03)

Elemental Analysis
Carbon, Hydrogen, Nitrogen and Sulfur

Trace Elements
Aluminium (Al), Boron (B), Calcium (Ca), Copper (Cu), Iron (Fe), Magnesium (Mg), Manganese (Mn), Phosphorous (P), Potassium (K), Sodium (Na, Zinc (Zn)

Matrix Group: Cereals, Bread, Rice and Maize

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service