Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92572

Supelco

Silica gel on TLC Al foils

L × W 5 cm × 20 cm, without fluorescence indicator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

material

aluminum support
silica gel 60 matrix

feature

binder Polymeric
fluorescent indicator: no

manufacturer/tradename

Sigma-Aldrich

technique(s)

thin layer chromatography (TLC): suitable

L × W

5 cm × 20 cm

layer thickness

200 μm

Looking for similar products? Visit Product Comparison Guide

General description

  • Outstanding wettability (even for 100% aqueous visualisation reagents)
  • Easy and precise cutting (no flaking of silica)
  • Excellent separation efficiency and reproducibility from batch to batch
Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation resulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is widely used as layer material for TLC. Plaster of Paris is generally used as binding agent to hold silica gel firmly to the TLC plates. Silica gel is applied as sorbent to aluminium foils.

Application

Silica gel on TLC Alu foils may be used to perform thin layer chromatography.

Other Notes

this product will be discontinued, please find the alternative product here :
105547

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph C. Touchstone
Practice of Thin Layer Chromatography (1992)
A simple method for production of pure silica from rice hull ash
Kalapathy, U., A. Proctor, and J. Shultz.
Bioresource Technology, 73, 257-262 (2000)
Silica gel supported zirconocene dichloride/methylalumoxane catalysts for ethylene polymerization: Effects of heterogenation on activity, polymer microstructure and product morphology.
Janiak, Christoph, and Bernhard Rieger.
Angew. Makromol. Chem., 215, 47-57 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service