Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91324

Supelco

N-Lauroylsarcosine sodium salt

analytical standard

Synonym(s):

N-Dodecanoyl-N-methylglycine sodium salt, Sarkosyl NL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CON(CH3)CH2COONa
CAS Number:
Molecular Weight:
293.38
Beilstein/REAXYS Number:
5322974
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

assay

≥97.0% (HPLC)

form

solid

shelf life

limited shelf life, expiry date on the label

mol wt

micellar avg mol wt 600

aggregation number

2

CMC

14.6 mM (20-25°C)

SMILES string

[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

InChI

1S/C15H29NO3.Na/c1-3-4-5-6-7-8-9-10-11-12-14(17)16(2)13-15(18)19;/h3-13H2,1-2H3,(H,18,19);/q;+1/p-1

InChI key

KSAVQLQVUXSOCR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Anionic surfactant.

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Eye Dam. 1 - Skin Irrit. 2

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service