Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90951

Supelco

Sodium palmitate

analytical standard

Synonym(s):

Hexadecanoic acid sodium salt, Palmitic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COONa
CAS Number:
Molecular Weight:
278.41
Beilstein/REAXYS Number:
3575882
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

assay

≥98.0% (GC)

form

powder

shelf life

limited shelf life, expiry date on the label

mp

283-290 °C (lit.)

Storage temp.

2-8°C

SMILES string

[Na+].CCCCCCCCCCCCCCCC([O-])=O

InChI

1S/C16H32O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18;/h2-15H2,1H3,(H,17,18);/q;+1/p-1

Inchi Key

GGXKEBACDBNFAF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service