Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90486

Supelco

Triethoxyvinylsilane

analytical standard

Synonym(s):

(Triethoxysilyl)ethylene, Vinyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHSi(OC2H5)3
CAS Number:
Molecular Weight:
190.31
Beilstein/REAXYS Number:
1767229
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

assay

≥98.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.398 (lit.)
n20/D 1.398-1.400

bp

160-161 °C (lit.)
62-63 °C/20 mmHg (lit.)

density

0.903 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](OCC)(OCC)C=C

InChI

1S/C8H18O3Si/c1-5-9-12(8-4,10-6-2)11-7-3/h8H,4-7H2,1-3H3

InChI key

FWDBOZPQNFPOLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

93.2 °F - closed cup

flash_point_c

34 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service