Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90480

Supelco

Octacosanoic acid

analytical standard

Synonym(s):

Montanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)26CO2H
CAS Number:
Molecular Weight:
424.74
Beilstein/REAXYS Number:
1801616
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

assay

≥97.5% (GC)

form

flakes

shelf life

limited shelf life, expiry date on the label

mp

91-93 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C28H56O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28(29)30/h2-27H2,1H3,(H,29,30)

InChI key

UTOPWMOLSKOLTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service