Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90129R

Supelco

Aroclor 1260 solution

1000 μg/mL in hexane, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

packaging

ampule of 1 mL

concentration

1000 μg/mL in hexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

room temp

InChI

1S/C12H4Cl6/c13-7-3-1-5(9(15)11(7)17)6-2-4-8(14)12(18)10(6)16/h1-4H

InChI key

BTAGRXWGMYTPBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aroclor 1260 solution may be used as a 5-point calibration standard for the determination of polychlorinated biphenyl in aroclor formulations using high resolution gas chromatography method coupled with electron capture detector (HRGC-ECD) or electrolytic conductivity detector (ELCD).[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-14.8 °F - closed cup

flash_point_c

-26 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Risk-Management Strategy for PCB-Contaminated Sediments (2001)
Chunlei Li et al.
Huan jing ke xue= Huanjing kexue, 23(2), 120-122 (2002-06-07)
The dosage of TiO2 and its surface-loaded silver had notable effect on the photocatalytic degradation of Aroclor1260, under ultraviolet light. The dosage of TiO2 had no proportion to the degradation rate of Aroclor1260, but had an optimal value, which was
Nebojsa L Andric et al.
Toxicology, 194(1-2), 65-75 (2003-11-26)
The mechanism by which Aroclors and other polychlorinated biphenyls (PCBs) inhibit testicular androgenesis in vivo and in vitro has not been characterized. Here we studied in adult rats the effects of intratesticular (i.t.t.), intraperitoneal (i.p.) and by gavage (p.o.) administration
Bianca Kuipers et al.
Canadian journal of microbiology, 49(1), 9-14 (2003-04-04)
We investigated the microbial reductive dechlorination of both weathered (aged) and nonweathered (freshly added) Aroclor 1260 in aerobic soil from Resolution Island, Nunavut, Canada. Initial polychlorinated biphenyl (PCB) concentrations were 106 and 100 ppm, respectively. The aerobic soil samples were
Brian A Mayes et al.
Regulatory toxicology and pharmacology : RTP, 35(3), 289-295 (2002-08-31)
Human health risk assessments involving contaminated soil include dermal absorption as a potential pathway contributing to the total exposure burden. For PCB-contaminated soil, the U.S. Environmental Protection Agency uses a dermal absorption factor of 14%, based on a 1993 study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service